1-(4-(Tert-pentyl)phenyl)propan-1-ol ≥97%

Supplier: AOBChem USA

Warning

355353-5G 355353-25G 355353-1G 355353-500MG
77632-912EA 1206 USD
77632-912 77658-908 77646-326 77650-322
1-(4-(Tert-pentyl)phenyl)propan-1-ol ≥97%
1-(4-(Tert-pentyl)phenyl)propan-1-ol

Precaución: For Research Use Only

Formula: 206.324 g/mol
MW: C₁₄H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD21731482
CAS Number: 1504114-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR