3-(5-Chloro-2-fluoro-3-methylphenyl)oxetan-3-ol ≥95%

Supplier: AOBChem USA

Warning

355291-5G 355291-1G 355291-25G 355291-500MG
77628-760EA 1484.23 USD
77628-760 77638-298 77658-906 77646-318
3-(5-Chloro-2-fluoro-3-methylphenyl)oxetan-3-ol ≥95%
3-(5-Chloro-2-fluoro-3-methylphenyl)oxetan-3-ol

Precaución: For Research Use Only

Formula: C₁₀H₁₀ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD35189670
CAS Number: 3002445-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR