2,7-Dimethyl-2H-indazole-5-boronic acid, pinacol ester ≥95%

Supplier: AOBChem USA

Warning

35525-5G 35525-1G 35525-500MG
77655-338EA 2237 USD
77655-338 77646-314 77646-316
2,7-Dimethyl-2H-indazole-5-boronic acid, pinacol ester ≥95%
2,7-Dimethyl-2H-indazole-5-boronic acid, pinacol ester

Precaución: For Research Use Only

Formula: C₁₅H₂₁BN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD12028668
CAS Number: 1310383-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR