3-(1,3-Dioxolan-2-yl)-5-(trifluoromethyl)-benzoic acid ≥95%

Supplier: AOBChem USA

Warning

35518-5G 35518-1G 35518-500MG
77628-758EA 1484.23 USD
77628-758 77642-310 77646-312
3-(1,3-Dioxolan-2-yl)-5-(trifluoromethyl)-benzoic acid ≥95%
3-(1,3-Dioxolan-2-yl)-5-(trifluoromethyl)-benzoic acid

Precaución: For Research Use Only

Formula: 262.182 g/mol
MW: C₁₁H₉F₃O₄
Storage Temperature: Ambient
MDL Number: MFCD31805805
CAS Number: 2271443-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR