2-Chloro-6-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

35504-5G 35504-25G
77634-912EA 61 USD
77634-912 77658-904
2-Chloro-6-methylbenzaldehyde ≥97%
2-Chloro-6-methylbenzaldehyde

Precaución: For Research Use Only

Formula: C₈H₇ClO
Storage Temperature: Ambient
MDL Number: MFCD01934409
CAS Number: 1194-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR