1-[4-(1-Naphthalenyl)phenyl]-1-propanone ≥95%

Supplier: AOBChem USA

Warning

354564-5G 354564-25G 354564-1G 354564-500MG
77628-756EA 1484.23 USD
77628-756 77655-334 77642-308 77646-310
1-[4-(1-Naphthalenyl)phenyl]-1-propanone ≥95%
1-[4-(1-Naphthalenyl)phenyl]-1-propanone

Precaución: For Research Use Only

Formula: 260.33 g/mol
MW: C₁₉H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD35189509
CAS Number: 1228926-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR