1-[4-(1-Naphthalenyl)phenyl]ethanone ≥95%

Supplier: AOBChem USA

Warning

354558-5G 354558-1G 354558-500MG 354558-25G
77628-754EA 1553.24 USD
77628-754 77638-294 77652-700 77655-332
1-[4-(1-Naphthalenyl)phenyl]ethanone ≥95%
1-[4-(1-Naphthalenyl)phenyl]ethanone

Precaución: For Research Use Only

Formula: 246.3 g/mol
MW: C₁₈H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD12859866
CAS Number: 216576-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR