(3,5-Dibromo-4-methylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

35445-5G 35445-1G 35445-25G 35445-500MG
77632-908EA 960 USD
77632-908 77638-292 77655-330 77646-308
(3,5-Dibromo-4-methylphenyl)methanol ≥95%
(3,5-Dibromo-4-methylphenyl)methanol

Precaución: For Research Use Only

Formula: 279.957 g/mol
MW: C₈H₈Br₂O
Storage Temperature: Ambient
MDL Number: MFCD12031828
CAS Number: 868860-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR