5-Bromo-2-fluoro-3-methoxybenzyl alcohol ≥95%

Supplier: AOBChem USA

Warning

35400-1G 35400-5G 35400-500MG
77638-290EA 719 USD
77638-290 77655-328 77650-318
5-Bromo-2-fluoro-3-methoxybenzyl alcohol ≥95%
5-Bromo-2-fluoro-3-methoxybenzyl alcohol

Precaución: For Research Use Only

Formula: C₈H₈BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD31805961
CAS Number: 1780693-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR