Ethyl 2,3-dibromo-6-fluorobenzoate ≥95%

Supplier: AOBChem USA

Warning

35398-5G 35398-1G 35398-500MG
77658-902EA 2384 USD
77658-902 77642-306 77646-306
Ethyl 2,3-dibromo-6-fluorobenzoate ≥95%
Ethyl 2,3-dibromo-6-fluorobenzoate

Precaución: For Research Use Only

Formula: C₉H₇Br₂FO₂
Storage Temperature: Ambient
MDL Number: MFCD28784656
CAS Number: 1805122-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR