Ethyl 2,6-dibromonicotinate ≥95%

Supplier: AOBChem USA

Warning

35388-5G 35388-1G 35388-25G 35388-500MG
77634-910EA 389 USD
77634-910 77638-286 77658-900 77646-302
Ethyl 2,6-dibromonicotinate ≥95%
Ethyl 2,6-dibromonicotinate

Precaución: For Research Use Only

Formula: 308.955 g/mol
MW: C₈H₇Br₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD00235042
CAS Number: 1214332-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR