1-Bromo-3-methoxy-2-propoxybenzene ≥95%

Supplier: AOBChem USA

Warning

35377-5G 35377-25G 35377-1G 35377-500MG
77632-904EA 798 USD
77632-904 77655-324 77642-304 77646-300
1-Bromo-3-methoxy-2-propoxybenzene ≥95%
1-Bromo-3-methoxy-2-propoxybenzene

Precaución: For Research Use Only

Formula: C₁₀H₁₃BrO₂
Storage Temperature: Ambient
MDL Number: MFCD31806016
CAS Number: 1864654-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR