2,3-Dibromobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

35374-5G 35374-1G 35374-25G 35374-500MG
77634-908EA 319 USD
77634-908 77638-284 77658-898 77650-316
2,3-Dibromobenzyl alcohol ≥97%
2,3-Dibromobenzyl alcohol

Precaución: For Research Use Only

Formula: C₇H₆Br₂O
Storage Temperature: Ambient
MDL Number: MFCD11520723
CAS Number: 1214331-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR