1-Bromo-2-(2-phenylethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

353623-5G 353623-25G 353623-1G 353623-500MG
77628-752EA 1484.23 USD
77628-752 77658-896 77642-302 77646-298
1-Bromo-2-(2-phenylethoxy)benzene ≥97%
1-Bromo-2-(2-phenylethoxy)benzene

Precaución: For Research Use Only

Formula: 277.16 g/mol
MW: C₁₄H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD11127636
CAS Number: 40515-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR