5-Bromo-2,3-difluoro-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

35358-5G 35358-1G 35358-25G 35358-500MG
77632-902EA 760 USD
77632-902 77638-282 77655-322 77650-314
5-Bromo-2,3-difluoro-4-methylbenzaldehyde ≥97%
5-Bromo-2,3-difluoro-4-methylbenzaldehyde

Precaución: For Research Use Only

Formula: C₈H₅BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD27992285
CAS Number: 1823871-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR