Cyclopropyl(2-fluoro-[1,1'-biphenyl]-4-yl)methanol ≥95%
Supplier: AOBChem USA
|
Warning
|
353345-5G
353345-25G
353345-1G
353345-500MG
77628-750EA
1648
USD
77628-750
77635-682
77642-300
77646-296
Cyclopropyl(2-fluoro-[1,1'-biphenyl]-4-yl)methanol ≥95%
Cyclopropyl(2-fluoro-[1,1'-biphenyl]-4-yl)methanol
Precaución: For Research Use Only
Formula:
242.288 g/mol MW: C₁₆H₁₅FO Storage Temperature: Ambient |
MDL Number:
MFCD35189309 CAS Number: 3002424-54-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...