1-([1,1'-Biphenyl]-3-yl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

353267-5G 353267-1G 353267-25G 353267-500MG
77628-748EA 1648 USD
77628-748 77638-278 77658-894 77646-294
1-([1,1'-Biphenyl]-3-yl)cyclopentanol ≥95%
1-([1,1'-Biphenyl]-3-yl)cyclopentanol

Precaución: For Research Use Only

Formula: 238.324 g/mol
MW: C₁₇H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD35189268
CAS Number: 3002528-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR