6-Bromo-4-fluoro-3-iodo-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

35295-5G 35295-25G 35295-1G 35295-500MG
77632-898EA 755 USD
77632-898 77655-314 77642-298 77650-310
6-Bromo-4-fluoro-3-iodo-1H-indazole ≥97%
6-Bromo-4-fluoro-3-iodo-1H-indazole

Precaución: For Research Use Only

Formula: C₇H₃BrFIN₂
Storage Temperature: Ambient
MDL Number: MFCD07781844
CAS Number: 887568-00-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR