2-Chloro-3-fluoro-4-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

35287-5G 35287-25G 35287-1G 35287-500MG
77632-894EA 983 USD
77632-894 77655-310 77646-288 77646-290
2-Chloro-3-fluoro-4-(methylthio)benzaldehyde ≥97%
2-Chloro-3-fluoro-4-(methylthio)benzaldehyde

Precaución: For Research Use Only

Formula: C₈H₆ClFOS
MW: 204.649 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32215273
CAS Number: 2271442-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR