1-Bromo-3-chloro-5-fluoro-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

35261-5G 35261-25G 35261-1G 35261-500MG
77632-888EA 1284 USD
77632-888 77655-302 77642-288 77650-296
1-Bromo-3-chloro-5-fluoro-2-methylbenzene ≥95%
1-Bromo-3-chloro-5-fluoro-2-methylbenzene

Precaución: For Research Use Only

Formula: C₇H₅BrClF
Storage Temperature: Ambient
MDL Number: MFCD28734045
CAS Number: 1780876-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR