2,6-Dibromobenzoic acid ethyl ester ≥97%
Supplier: AOBChem USA
|
Warning
|
35260-5G
35260-1G
35260-25G
35260-500MG
77632-886EA
924
USD
77632-886
77638-272
77655-300
77646-284
2,6-Dibromobenzoic acid ethyl ester ≥97%
2,6-Dibromobenzoic acid ethyl ester
Precaución: For Research Use Only
Formula:
307.967 g/mol MW: C₉H₈Br₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD11616905 CAS Number: 1214375-69-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...