4-(2-Methoxypyrimidin-5-yl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

35258-5G 35258-25G 35258-1G 35258-500MG
77632-884EA 1430 USD
77632-884 77655-296 77642-284 77650-292
4-(2-Methoxypyrimidin-5-yl)benzaldehyde ≥95%
4-(2-Methoxypyrimidin-5-yl)benzaldehyde

Precaución: For Research Use Only

Formula: C₁₂H₁₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD31805962
CAS Number: 1785548-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR