5-Bromo-2-methoxy-3-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

35249-5G 35249-1G 35249-25G 35249-500MG
77634-904EA 536 USD
77634-904 77638-270 77655-294 77650-290
5-Bromo-2-methoxy-3-(methylthio)pyridine ≥97%
5-Bromo-2-methoxy-3-(methylthio)pyridine

Precaución: For Research Use Only

Formula: C₇H₈BrNOS
MW: 234.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28060209
CAS Number: 1780085-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR