5-Bromo-2-ethoxy-1,3-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

35216-5G 35216-1G 35216-25G 35216-500MG
77632-882EA 1319 USD
77632-882 77638-268 77658-890 77646-282
5-Bromo-2-ethoxy-1,3-dimethylbenzene ≥97%
5-Bromo-2-ethoxy-1,3-dimethylbenzene

Precaución: For Research Use Only

Formula: C₁₀H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD18917790
CAS Number: 126209-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR