2-Fluoro-4-(trifluoromethoxy)benzyl alcohol ≥95%

Supplier: AOBChem USA

Warning

351709-5G 351709-1G 351709-500MG 351709-25G
77634-900EA 346 USD
77634-900 77638-266 77652-698 77658-888
2-Fluoro-4-(trifluoromethoxy)benzyl alcohol ≥95%
2-Fluoro-4-(trifluoromethoxy)benzyl alcohol

Precaución: For Research Use Only

Formula: C₈H₆F₄O₂
MW: 210.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16652441
CAS Number: 1240257-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR