2-(Benzyloxy)-3-bromobenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
35157-5G
35157-1G
35157-25G
35157-500MG
77632-878EA
1207
USD
77632-878
77638-262
77655-288
77650-286
2-(Benzyloxy)-3-bromobenzoic acid ≥97%
2-(Benzyloxy)-3-bromobenzoic acid
Precaución: For Research Use Only
Formula:
307.139 g/mol MW: C₁₄H₁₁BrO₃ Storage Temperature: Ambient |
MDL Number:
MFCD13479730 CAS Number: 1293226-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...