3-Bromo-2-ethoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

35116-5G 35116-25G 35116-1G 35116-500MG
77632-874EA 1100 USD
77632-874 77655-284 77642-278 77646-276
3-Bromo-2-ethoxybenzoic acid ≥95%
3-Bromo-2-ethoxybenzoic acid

Precaución: For Research Use Only

Formula: C₉H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD14695223
CAS Number: 1275532-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR