1-[6-(4-Fluorophenyl)-2-methyl-3-pyridinyl]ethanone ≥95%

Supplier: AOBChem USA

Warning

351088-5G 351088-1G 351088-500MG
77658-884EA 2304 USD
77658-884 77642-276 77650-282
1-[6-(4-Fluorophenyl)-2-methyl-3-pyridinyl]ethanone ≥95%
1-[6-(4-Fluorophenyl)-2-methyl-3-pyridinyl]ethanone

Precaución: For Research Use Only

Formula: 229.25 g/mol
MW: C₁₄H₁₂FNO
Storage Temperature: Ambient
MDL Number: MFCD08059534
CAS Number: 914635-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR