7-Methyl-1H-indazole-5-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

35049-5G 35049-1G 35049-500MG
77658-882EA 2770 USD
77658-882 77642-274 77646-270
7-Methyl-1H-indazole-5-carbaldehyde ≥95%

Precaución: For Research Use Only

Formula: C₉H₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD16619770
CAS Number: 635712-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR