3,6-Dichloro-2-fluoro-α-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

350344-500MG 350344-5G 350344-1G
77652-696EA 379 USD
77652-696 77658-880 77642-270
3,6-Dichloro-2-fluoro-α-methylbenzenemethanol ≥95%
3,6-Dichloro-2-fluoro-α-methylbenzenemethanol

Precaución: For Research Use Only

Formula: 209.05 g/mol
MW: C₈H₇Cl₂FO
Storage Temperature: Ambient
MDL Number: MFCD28528424
CAS Number: 1780393-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR