1-(Cyclopentyloxy)-2-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

349944-1G 349944-5G 349944-500MG
77634-896EA 1001 USD
77634-896 77658-878 77646-268
1-(Cyclopentyloxy)-2-nitrobenzene ≥97%
1-(Cyclopentyloxy)-2-nitrobenzene

Precaución: For Research Use Only

Formula: 207.23 g/mol
MW: C₁₁H₁₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD13689282
CAS Number: 29026-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR