3-(2,2,2-Trifluoroacetyl)-benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

34980-5G 34980-25G 34980-1G 34980-500MG
77634-894EA 587 USD
77634-894 77655-278 77642-268 77650-276
3-(2,2,2-Trifluoroacetyl)-benzaldehyde ≥97%
3-(2,2,2-Trifluoroacetyl)-benzaldehyde

Precaución: For Research Use Only

Formula: 202.13 g/mol
MW: C₉H₅F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD34755976
CAS Number: 1830341-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR