5-Bromo-2-(2,5-dimethyl-1H-pyrrol-1-yl)pyridine ≥95%

Supplier: AOBChem USA

Warning

34947-5G 34947-25G
77634-892EA 114 USD
77634-892 77658-876
5-Bromo-2-(2,5-dimethyl-1H-pyrrol-1-yl)pyridine ≥95%
5-Bromo-2-(2,5-dimethyl-1H-pyrrol-1-yl)pyridine

Precaución: For Research Use Only

Formula: C₁₁H₁₁BrN₂
MW: 251.1200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01125286
CAS Number: 228710-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR