2-Bromo-4-fluoro-3-methyl-1-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

34894-1G 34894-5G 34894-500MG
77634-888EA 781 USD
77634-888 77658-872 77646-266
2-Bromo-4-fluoro-3-methyl-1-(trifluoromethyl)benzene ≥95%
2-Bromo-4-fluoro-3-methyl-1-(trifluoromethyl)benzene

Precaución: For Research Use Only

Formula: C₈H₅BrF₄
MW: 257.023 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631807
CAS Number: 2167902-49-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR