Cyclopropyl(4'-propyl[1,1'-biphenyl]-4-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

348483-5G 348483-1G 348483-500MG
77658-870EA 2001 USD
77658-870 77642-260 77646-258
Cyclopropyl(4'-propyl[1,1'-biphenyl]-4-yl)methanone ≥97%
Cyclopropyl(4'-propyl[1,1'-biphenyl]-4-yl)methanone

Precaución: For Research Use Only

Formula: C₁₉H₂₀O
Storage Temperature: Ambient
MDL Number: MFCD28409698
CAS Number: 2173094-91-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR