2,2,2-Trifluoro-1-(4'-propyl[1,1'-biphenyl]-4-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

348414-5G 348414-1G 348414-500MG
77655-270EA 2356 USD
77655-270 77642-258 77650-272
2,2,2-Trifluoro-1-(4'-propyl[1,1'-biphenyl]-4-yl)ethanone ≥97%
2,2,2-Trifluoro-1-(4'-propyl[1,1'-biphenyl]-4-yl)ethanone

Precaución: For Research Use Only

Formula: 292.3 g/mol
MW: C₁₇H₁₅F₃O
Storage Temperature: Ambient
MDL Number: MFCD07783576
CAS Number: 1256481-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR