1-(4'-Propyl[1,1'-biphenyl]-4-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

348409-5G 348409-1G 348409-500MG
77658-868EA 2001 USD
77658-868 77642-256 77650-270
1-(4'-Propyl[1,1'-biphenyl]-4-yl)ethanone ≥97%
1-(4'-Propyl[1,1'-biphenyl]-4-yl)ethanone

Precaución: For Research Use Only

Formula: 238.32 g/mol
MW: C₁₇H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD00859438
CAS Number: 60137-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR