2,6-Diethoxyisonicotinic acid ≥97%

Supplier: AOBChem USA

Warning

34834-5G 34834-25G 34834-1G 34834-500MG
77632-868EA 1121 USD
77632-868 77658-866 77642-254 77650-268
2,6-Diethoxyisonicotinic acid ≥97%
2,6-Diethoxyisonicotinic acid

Precaución: For Research Use Only

Formula: 211.215 g/mol
MW: C₁₀H₁₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD03086064
CAS Number: 5397-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR