6-Bromoquinoline-N-oxide ≥97%

Supplier: AOBChem USA

Warning

34832-5G 34832-1G 34832-500MG 34832-25G
77634-886EA 346 USD
77634-886 77638-254 77652-694 77655-268
6-Bromoquinoline-N-oxide ≥97%
6-Bromoquinoline-N-oxide

Precaución: For Research Use Only

Formula: 224.054 g/mol
MW: C₉H₆BrNO
Storage Temperature: Ambient
MDL Number: MFCD08062621
CAS Number: 6563-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR