4-Bromothiazole-5-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

34700-1G 34700-5G 34700-500MG
77638-246EA 505 USD
77638-246 77655-258 77646-244
4-Bromothiazole-5-carboxylic acid ≥97%
4-Bromothiazole-5-carboxylic acid

Precaución: For Research Use Only

Formula: 208.033 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11111596
CAS Number: 1244949-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR