(4-(Dimethylamino)-2-fluorophenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

346771-5G 346771-1G 346771-500MG
77628-730EA 1484.23 USD
77628-730 77646-242 77650-264
(4-(Dimethylamino)-2-fluorophenyl)boronic acid ≥97%
(4-(Dimethylamino)-2-fluorophenyl)boronic acid

Precaución: For Research Use Only

Formula: 182.988 g/mol
MW: C₈H₁₁BFNO₂
Storage Temperature: Ambient
MDL Number: MFCD31638891
CAS Number: 1857375-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR