2-(3-Chloro-2-fluoropyridin-4-yl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

346634-5G 346634-1G 346634-25G 346634-500MG
77628-728EA 1718.04 USD
77628-728 77638-244 77655-256 77650-262
2-(3-Chloro-2-fluoropyridin-4-yl)propan-2-ol ≥95%
2-(3-Chloro-2-fluoropyridin-4-yl)propan-2-ol

Precaución: For Research Use Only

Formula: C₈H₉ClFNO
Storage Temperature: Ambient
MDL Number: MFCD35187240
CAS Number: 3002531-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR