5-Bromo-1-chloro-2-fluoro-3-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

34660-5G 34660-25G 34660-1G
77634-882EA 223 USD
77634-882 77655-254 77642-250
5-Bromo-1-chloro-2-fluoro-3-iodobenzene ≥97%
5-Bromo-1-chloro-2-fluoro-3-iodobenzene

Precaución: For Research Use Only

Formula: C₆H₂BrClFI
MW: 335.34 g/mol
Storage Temperature: Ambient
CAS Number: 56141-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR