2,3-Difluoro-4-iodo-5-(trifluoromethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

346343-5G 346343-1G 346343-25G 346343-500MG
77628-726EA 1839.58 USD
77628-726 77638-242 77658-864 77650-260
2,3-Difluoro-4-iodo-5-(trifluoromethyl)pyridine ≥97%
2,3-Difluoro-4-iodo-5-(trifluoromethyl)pyridine

Precaución: For Research Use Only

Formula: 308.975 g/mol
MW: C₆HF₅IN
Storage Temperature: Ambient
CAS Number: 3031785-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR