2-Bromo-3-phenylthiophene ≥97%

Supplier: AOBChem USA

Warning

34561-5G 34561-1G 34561-500MG
77655-250EA 2053 USD
77655-250 77642-248 77650-258
2-Bromo-3-phenylthiophene ≥97%
2-Bromo-3-phenylthiophene

Precaución: For Research Use Only

Formula: 239.132 g/mol
MW: C₁₀H₇BrS
Storage Temperature: Ambient
MDL Number: MFCD16618639
CAS Number: 10341-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR