4-(5-Bromopyridin-2-yl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

34557-5G 34557-1G 34557-25G 34557-500MG
77632-864EA 1207 USD
77632-864 77638-240 77655-248 77650-256
4-(5-Bromopyridin-2-yl)benzaldehyde ≥97%
4-(5-Bromopyridin-2-yl)benzaldehyde

Precaución: For Research Use Only

Formula: C₁₂H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD11227191
CAS Number: 910547-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR