1,1-Dimethylethyl 2-bromobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

344946-5G 344946-1G 344946-25G 344946-500MG
77634-876EA 588 USD
77634-876 77638-238 77655-242 77650-250
1,1-Dimethylethyl 2-bromobenzeneacetate ≥95%
1,1-Dimethylethyl 2-bromobenzeneacetate

Precaución: For Research Use Only

Formula: 271.15 g/mol
MW: C₁₂H₁₅BrO₂
Storage Temperature: Ambient
MDL Number: MFCD11976801
CAS Number: 123088-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR