4-Bromo-5-fluoro-2-methylbenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

34492-5G 34492-1G 34492-25G 34492-500MG
77634-874EA 536 USD
77634-874 77638-236 77658-858 77646-238
4-Bromo-5-fluoro-2-methylbenzyl alcohol ≥97%
4-Bromo-5-fluoro-2-methylbenzyl alcohol

Precaución: For Research Use Only

Formula: C₈H₈BrFO
MW: 219.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22552984
CAS Number: 1352899-41-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR