3,5-Difluoro-2-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

34472-5G 34472-1G 34472-500MG
77655-238EA 2203 USD
77655-238 77642-238 77646-236
3,5-Difluoro-2-(methylthio)benzoic acid ≥97%
3,5-Difluoro-2-(methylthio)benzoic acid

Precaución: For Research Use Only

Formula: C₈H₆F₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD34755964
CAS Number: 2807444-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR