3-Bromo-5-fluoro-2-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

34465-5G 34465-1G 34465-500MG
77628-722EA 1484.23 USD
77628-722 77642-236 77646-234
3-Bromo-5-fluoro-2-methylbenzaldehyde ≥95%
3-Bromo-5-fluoro-2-methylbenzaldehyde

Precaución: For Research Use Only

Formula: C₈H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD19440366
CAS Number: 1378962-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR